Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL20RB Mouse anti-Human, Clone: 1A2A3, Proteintech
SDP

Catalog No. 501736322 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6322 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6322 Supplier Proteintech Group Inc Supplier No. 602211IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

IL20RB and IL20RA.[supplied by OMIM, Feb 2009].

Specifications

Antigen IL20RB
Applications Western Blot
Classification Monoclonal
Clone 1A2A3
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL20RB
Gene Accession No. Q6UXL0
Gene Alias DIRS1, FNDC6, IL 20 receptor subunit beta, IL 20R beta, IL 20R2, IL 20RB, IL20RB, interleukin 20 receptor beta
Gene Symbols IL20RB
Host Species Mouse
Immunogen IL20RB Fusion Protein Ag14368
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53833
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.