Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL21R Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675541
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-541 Supplier Invitrogen™ Supplier No. PA141412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse samples based on sequence homology. Suggested positive control: Raji whole cell lysate, NIH-3T3 whole cell lysate. Immunogen sequence: CSLHRSAHNA THATYT & CDFTSPGDEG PPRSYLR.

The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL21R
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene Il21r
Gene Accession No. Q9HBE5
Gene Alias CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273
Gene Symbols Il21r
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 65-80 and 505-521 of human IL-21R.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50615
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.