Learn More
ACROBiosystems Recombinant Protein;Human IL-21 R Protein, Fc Tag;HEK293;200UG;ILR,IL21R,CD360,NILR,IL-21R
Supplier: ACROBiosystems ILRH5257200UG
Recombinant Protein;Human IL-21 R Protein, Fc Tag;200UG;Human IL-21 R, Fc Tag (ILR-H5257) is expressed from human 293 cells (HEK293). It contains AA Cys 20 - Pro 236 (Accession # AAH04348).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.6 kDa. As a result of glycosylation, the protein migrates as 65-80 kDa under reducing (R) condition, and 130-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Biotinylated Human IL-21, Fc Tag (Cat. No. IL1-H82F7) at 2 μg/mL (100 μL/well) can bind Human IL-21 R, Fc Tag (Cat. No. ILR-H5257) with a linear range of 2-31 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.