Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL28B/ IFNL3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734258 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4258 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4258 Supplier Proteintech Group Inc Supplier No. 241991AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. As new cytokines are molecularly characterized, they are assigned an IL number to maintain a standard nomenclature. The interleukins are secreted by immune cells (mainly macrophages, B-cells or T-cells) that regulate a wide range of immune system functions. The functions of different interleukins vary from the regulation of inflammatory and immune responses to the regulation of other interleukins. IL-28 and IL-29 are induced by viral infection and show antiviral activity. IL-28 may play a role in antiviral immunity through up-regulation of MHC class I antigen expression. IL-28 is a ligand for the heterodimeric class II cytokine receptor composed of IL10RB and IL28RA.

Specifications

Antigen IL28B/ IFNL3
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFNL3
Gene Accession No. Q8IZI9
Gene Alias Cytokine Zcyto22, IFN lambda 3, IFN lambda 4, IFNL3, IFNL4, IL 28B, IL 28C, IL28B, IL28C, Interferon lambda 3, Interferon lambda 4, Interleukin 28B, Interleukin 28C, ZCYTO22
Gene Symbols IFNL3
Host Species Rabbit
Immunogen IL28B/ IFNL3 Fusion Protein Ag21491
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.