Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL31RA Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-200128798
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119229 200 μL
PIPA5121824 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119229 Supplier Invitrogen™ Supplier No. PA5119229
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL31RA Polyclonal Antibody for Western Blot, ICC/IF

IL-31 is a T cell cytokine that is preferentially produced by T helper type 2 cells. IL-31 signals through a heterodimeric receptor composed of the IL-31 receptor (IL-31R) and the oncostatin M receptor (OSM). This receptor complex recruits JAK1, JAK2, Stat1, Stat3 and Stat5 signaling pathways, as well as the PI3 kinase/AKT cascade. SHP-2 and Shc adapter molecules are also recruited and contribute to an increased activation of the MAP kinase pathway in response to IL-31. Overexpression of IL-31 in mice results in pruritus and skin dermatitis resembling human atopic dermatitis (AD). Comparisons between skin from patients with AD and healthy skin showed IL-31R expression at higher levels on epidermal keratinocytes in AD samples. Infiltrating cells, more numerous in skin from patients with AD compared with that of healthy individuals, expressed IL-31 mRNA. IL-31 may participate in the cause of itch sensation and promote scratching behavior in NC/Nga mice with atopic dermatitis, and may represent a novel target for antipruritic drug development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL31RA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IL31RA
Gene Accession No. Q8NI17
Gene Alias class I cytokine receptor; CRL; CRL 3; CRL3; CRL3Glmr; cytokine receptor NR10; cytokine receptor-like 3; GLM R; Glmr; GLM-R; GLM-RMGC125346; GP130 like receptor; gp130-like monocyte receptor; Gp130-like receptor; GPL; HGLM R; HGLMR; hGLM-R; IL 31RA; IL-31 receptor subunit alpha; IL31R subunit alpha; IL-31R subunit alpha; Il31ra; IL-31RA; IL-31RAGPLCRL; IL-31R-alpha; interleukin 31 receptor A; interleukin 31RA; interleukin-31 receptor subunit alpha; MGC125346; mGLM-R; novel cytokine receptor 10; NR10; PLCA 2; PLCA2; PRO21384; soluble type I cytokine receptor CRL3; UNQ6368/PRO21073/PRO21384; ZcytoR17
Gene Symbols IL31RA
Host Species Rabbit
Immunogen Recombinant protein Interleukin 31 Receptor A. The antigen corresponds to amino acid range 205-456 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100522044, 133396
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.