Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL36RN Mouse anti-Human, Clone: 5F11G6, Proteintech
SDP

Catalog No. 501736393 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6393 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6393 Supplier Proteintech Group Inc Supplier No. 602981IG150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.

Specifications

Antigen IL36RN
Applications Western Blot
Classification Monoclonal
Clone 5F11G6
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IL36RN
Gene Accession No. Q9UBH0
Gene Alias FIL1, FIL1 delta, FIL1(DELTA), FIL1D, IL 1 delta, IL 1 related protein 3, IL 1F5, IL 1HY1, IL 1L1, IL 1ra homolog 1, IL 1RP3, IL1F5, IL1HY1, IL1L1, IL1RP3, Interleukin 1 delta, Interleukin 1 family member 5, Interleukin 1 HY1, Interleukin 1 like protein 1
Gene Symbols IL36RN
Host Species Mouse
Immunogen IL36RN Fusion Protein Ag3780
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26525
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.