Learn More
ACROBiosystems Recombinant Protein;Human IL-37 / IL-1F7b Protein;E.coli;50UG;ILB,IL1F7,IL37,FIL1Z,IL1H4,IL1RP1,IL-1F7,IL-37,Interleukin-37,Interleukin 37,IL-1H4,IL-1F7b,IL1F7b
Supplier: ACROBiosystems ILBH514150UG
Recombinant Protein;Human IL-37 / IL-1F7b Protein;50UG;Human IL-37, His Tag (ILB-H5141) is expressed from E.coli cells. It contains AA Val 46 - Asp 218 (Accession # AAH20637).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 20.3 kDa. The protein migrates as 20.3 kDa under reducing (R) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.