Learn More
Sino Biological Human IL3RA / CD123 Protein (His Tag) 100ug
Supplier:  Sino Biological 10518H08H100
Encompass_Preferred
A DNA sequence encoding the human IL3RA (NP_002174.1) extracellular domain (Met1-Arg305) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-6297
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.