Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL4I1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113266 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113266 Supplier Invitrogen™ Supplier No. PA5113266
  • $541.00
    $447.65 / Each of 1

    Save $93.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IL4I1 Polyclonal Antibody for ELISA, Western Blot

This gene encodes a protein with limited similarity to L-amino acid oxidase which contains the conserved amino acids thought to be involved in catalysis and binding of flavin adenine dinucleotide (FAD) cofactor. The expression of this gene can be induced by interleukin 4 in B cells, however, expression of transcripts containing the first two exons of the upstream gene is found in other cell types. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL4I1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IL4I1
Gene Accession No. Q96RQ9
Gene Alias FIG1; Fig-1 protein; Fig1-ps; hFIG1; IL4I1; IL4I1_2; IL4-induced protein 1; interleukin 4 induced 1; interleukin four induced 1; interleukin-4-induced protein 1; interleukin-four induced gene 1; interleukin-four induced gene-1 protein; interleukin-four induced protein-1; LAAO; L-amino acid oxidase; L-amino-acid oxidase; LAO; LOC100360621; mFIG1; Protein Fig-1; rCG53598-like; UNQ636/PRO1265
Gene Symbols IL4I1
Host Species Rabbit
Immunogen Recombinant Human L-amino-acid oxidase protein (139-339AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259307
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.