Learn More
Invitrogen™ IL5RA Monoclonal Antibody (26815), APC

Description
In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) IL-5 R beta, rhIL-9 R, rhIL-4 R, rmIL-5 R alpha, or rhIL-13 R alpha 1.
Specifications
Specifications
| Antigen | IL5RA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 26815 |
| Conjugate | APC |
| Formulation | Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide |
| Gene | IL5RA |
| Gene Accession No. | Q01344 |
| Gene Alias | CD125; CD125 antigen; CDw125; HSIL5R3; IL-5 receptor alpha chain; IL-5 receptor subunit alpha; Il5r; IL-5R subunit alpha; Il5ra; IL-5RA; IL-5R-alpha; interleukin 5 receptor subunit alpha; interleukin 5 receptor type 3; interleukin 5 receptor, alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor subunit alpha |
| Gene Symbols | IL5RA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.