Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL6R Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™
SDP

Catalog No. 501664486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-4486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-4486 Supplier OriGene Supplier No. CF506855
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

IL6R (interleukin-6 receptor subunit alpha) is a part of the receptor for interleukin 6. The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signal transducing component. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.

Specifications

Antigen IL6R
Applications Western Blot
Classification Monoclonal
Clone OTI1B5
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene IL6R
Gene Accession No. P08887
Gene Alias CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
Gene Symbols IL6R
Host Species Mouse
Immunogen Full length human recombit protein of human IL6R produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.