Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover CBRN Cartridge Filters, Set of 3

Catalog No. 19631034
Change view
Click to view available options
CBRN filter cartridges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-631-034 CBRN filter cartridges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-631-034 Supplier ILC Dover Supplier No. S2016
Add to Cart
Add to Cart

Easy to use twist and lock cartridge filters that come in a hermetically sealed foil pouch. NIOSH approved, designed to deliver a CAP 2 protection rating. Superior protection against military warfare agents, biologic threats, industrial chemical threats,and nuclear particulate threats.

  • Designed to deliver a CAP 2 protection rating
  • Superior protection against military warfare agents, biologic threats, industrial chemical threats, and nuclear particulate threats

Specifications

Battery Type NiMH
Cartridge Type CBRN Cartridge
Certifications/Compliance NIOSH
Color Black
Product Type Cartridge/Filter Combination
Format Universal
For Use With (Equipment) Cbrn system
Includes 3 CBRN cartridges
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.