ILC Dover SCape™ Air Purifying Escape Hoods

Delivers reliable protection with minimal training

Manufacturer: ILC DOVER INC 60001130001

 View more versions of this product

Catalog No. 19-631-056

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH
    For Use With (Application) Provides both carbon monoxide and smoke protection
    Includes Intergral 64 lmp blower with indicator light, exhaust valve, one-size neck seal, military grade filter canister, hepa filter, multi layer nylon film lens, battery, clear hood with large 360 vision
    Material Polyethylene
    Shelf Life 5 Years
    Size Universal
    Product Type Air Purifying Escape Hood

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit