Learn More
Mopec™ Illuminated Tissue Flotation Bath Lid
SDP
Catalog No. 22445790 Shop All Mopec Products
Change view
Click to view available options
Type:
Flotation Bath Lid
Catalog No. Type
22-445-790 Flotation Bath Lid
1 options
Catalog No. 22-445-790 Supplier Mopec™ Supplier No. BK200L
Add to Cart
Add to Cart
Illuminated Tissue Flotation Bath features an LED light and high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water. The LED light is dimmer controlled and can be switched between white or blue.
  • Lid for use with the Mopec™ Illuminated Tissue Flotation Bath
Specifications
Temperature Range 30°C to 70°C
Type Flotation Bath Lid
Electrical Requirements None