Learn More
Mopec™ Illuminated Tissue Flotation Bath and Slide Drying Work Station Bundle
SDP
Catalog No. 22445815 Shop All Mopec Products
Change view
Click to view available options
Electrical Requirements:
None
Catalog No. Electrical Requirements
22-445-815 None
1 options
Catalog No. 22-445-815 Supplier Mopec™ Supplier No. BK250
Add to Cart
Add to Cart
Illuminated Tissue Flotation Bath features an LED light and high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water.
  • Designed to take up minimal bench-top space
  • Slide Drying Mini Workstation fits directly over Illuminated Tissue Flotation Bath
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Temperature Range 30°C to 70°C
Type Deluxe Lighted Tissue Flotation Bath
Electrical Requirements None