Learn More
Mopec™ Illuminated Tissue Flotation Bath
SDP
Catalog No. 22445421 Shop All Mopec Products
Change view
Click to view available options
Electrical Requirements:
None
Catalog No. Electrical Requirements
22-445-421 None
1 options
Catalog No. 22-445-421 Supplier Mopec™ Supplier No. BK200
Add to Cart
Add to Cart
Illuminated Tissue Flotation Bath features an LED light and high-contrast background that allows users to better view tissue sections while also detecting foreign particles floating on the surface of the water. The LED light is dimmer controlled and can be switched between white or blue.
  • Designed to take up minimal bench-top space
  • Switch between blue and white LED back lighting with dimmer control for optimal viewing
  • Extra deep bowl to assist with tissue pick-up at any angle
  • Digital temperature control
  • Rapid heating system ensures system is ready to use in less time
  • Convenient slide drying area at the rear of the bath
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Optional lid helps improve temperature control and prevent surface particles
Specifications
Temperature Range 30°C to 70°C
Type Deluxe Lighted Tissue Flotation Bath
Electrical Requirements None