Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva illustra AutoSeq G-50
GSA_VA

Catalog No. 45002409 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-409 1000 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-002-409 Supplier Cytiva Supplier No. 27534003
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Remove fluorescent dye-terminators from sequencing reactions prior to analysis in an automated sequencer. Cytiva illustra AutoSeq G-50 columns are pre-filled with Sephadex G-50 for effective purification to ensure high-quality sequencing results.

illustra AutoSeq G-50 columns are specifically designed to remove residual fluorescent dye-terminators that can obscure data in automated sequencing. illustra AutoSeq G-50 consists of MicroSpin columns containing Sephadex G-50, pre-equilibrated in double-distilled water. This is important because even the small amount of salt in traditional buffers (such as TE buffer) can cause electrophoretic artifacts on salt-sensitive automated sequencers.
  • For rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing
  • Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
  • Columns that are ready to use enable sample application to collection of purified product in less than 4 minutes
  • Can also be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Column
Isolation Technology Spin Column
Product Type Purification Column
Quantity 1000 Columns
Sample Volume 12 to 25μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.