Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva blood genomicPrep Mini Spin Kit

Catalog No. 45001496 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-496 50 Preps
45-001-497 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-496 Supplier Cytiva Supplier No. 28904264
Add to Cart
Add to Cart

Includes

  • Spin columns with a novel silica membrane, lyophilized proteinase K powder, lysis solution, wash and elution buffers, microcentrifuge
  • collection tubes, a full protocol booklet, and a detachable, quick reference protocol card.

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from whole blood, buffy coat, bone marrow and nucleated red blood cells.

The purification process involves minimal shearing with typical yields of 4 to 6 μg of genomic DNA per 200μL sample with volumes accommodated up to 1mL. The method uses chaotropic agents to extract DNA from blood cells, denature protein components, and promote the selective binding of DNA to a novel silica-membrane. The elution volume can be modified as necessary for most downstream molecular biological applications.

  • High-quality genomic DNA with a greater proportion of higher molecular weight DNA and less shearing
  • Demonstrated application to various types of whole blood including human, horse, rabbit, rat and mouse
  • Increased ease of use with reduced pipetting volume changes, one centrifugation setting, and color-coded kit components
  • Validated in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests

TRUSTED_SUSTAINABILITY

Specifications

Description illustra blood genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from whole blood, buffy coat, bone marrow, and nucleated red blood cells.
For Use With (Application) Used for several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digestsSuitable for use in molecular biology applications including standard, multiplex and quantitative PCR; cloning; haplotyping; rest
Includes Spin columns prepacked with a novel Silica membrane, Lyophilized Proteinase K powder, Lysis solution, wash and Elution Buffers, microcentrifuge collection tubes, a full protocol booklet and a detachable, quick reference protocol card
No. of Reactions 50 Preps
Product Type illustra blood genomicPrep Mini Spin Kit
Purity 1.7
Sample Volume 50 μl whole blood, Buffy coat, bone marrow cells, 2 μl nucleated blood†, 300 μl whole blood‡
Test Time < 20 min
Yield 4-6 μg/200 μl whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.