Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva illustra ExoProStar 1-Step

Catalog No. 45200500 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
20
100
500
2000
5000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-200-500 20
45-200-501 100
45-200-502 500
45-200-503 2000
45-200-504 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-200-500 Supplier Cytiva Supplier No. US77701
Add to Cart
Add to Cart

Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. Comes in a single tube for maximum convenience.

illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!

The ExoProStar 1-Step PCR and Sequence Reaction Clean-Up reagent provides a simple, single-step method for removing unincorporated primers and nucleotides prior to downstream analysis. illustra Exonuclease I and Alkaline Phosphatase enzymes are combined in a single, stable mix requiring just one pipetting step to set up the clean-up reaction.

illustra ExoProStar 1-Step is based on GE™ founding patents for the clean-up of PCR reactions using Exonuclease 1 and Alkaline Phosphatase. This 1-step version is not available in USA, Canada and Japan. Instead, we offer a version with the enzymes in two separate tubes, the 29002090, and the faster version, 29046321.

TRUSTED_SUSTAINABILITY

Specifications

Format Tube
For Use With (Application) Enzymatic PCR clean up
No. of Reactions 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.