Learn More
Cytiva ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Catalog No. 45002352 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
500
Catalog No. No. of Reactions
45-002-352 500
1 options
Catalog No. 45-002-352 Supplier Cytiva Supplier No. US79050
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
  • Easy to automate
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Complete heat inactivation of the enzymes within 10 minutes
  • Fast 15 minute protocol
  • Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
  • Improves digestion efficiency with no degradation of the target PCR product
  • Sample is completely intact and ready for immediate use in manual or automated processes
TRUSTED_SUSTAINABILITY
Specifications
Description Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.
Isolation Technology Enzymatic PCR Clean-up Technology
For Use With (Application) PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
No. of Reactions 500
Product Type illustra ExoProStar S