Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GenomiPhi V2 DNA Amplification Kit

Catalog No. 45001223 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
100 reactions
25 reactions
500 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-223 500 reactions
45-001-221 25 reactions
45-001-222 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-223 Supplier Cytiva Supplier No. 25660032
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote

Includes

Sample buffer, Reaction buffer, Enzyme mix, and Lambda control DNA (10ng/μL)

Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.

  • Starting material can be purified DNA or nonpurified cell lysates
  • Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
  • Less hands-on time compared to traditional isolation methods
  • Representative amplification of the whole genome
  • Automation-friendly protocol with great reproducibility
  • High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
  • Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling.
Description Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample.
For Use With (Application) Genomic DNA Amplification
Includes 5 x 0.9mL Sample Buffer, 5 x 0.9mL Reaction Buffer, 5 x 100μL Enzyme mix, 20μL Control DNA (lambda)
No. of Reactions 500 reactions
Product Type illustra GenomiPhi V2 DNA Amplification Kit
Size > 10 kb
Test Time 1.5-2 h
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.