Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GFX 96 PCR Purification Kit
GSA_VA

Catalog No. 45001488 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
96-Well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-488 96-Well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-488 Supplier Cytiva Supplier No. 28903445
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Binding plates; Wash plates; Collection plates and color-coded bottles of capture buffer; Wash buffer; Choice of two elution buffers (Tris-HCl and sterile water); Instruction booklet

Simultaneously purify up to 96 PCR products of between 100 bp and 10 kbp in as little as 15 minutes

  • High yields of pure DNA recovered in a small volume of water or a low-ionic strength buffer
  • Typical recoveries are > 85% for PCR products of 100 bp to 10kb length; Salt removal > 99%
  • Prepacked glass-fiber matrix 96-well plates are validated for use with the Macherey Nagel NucleoVac 96-vacuum manifold
  • No need for ethanol precipitations and hazardous organic extractions

Fluorescent sequencing, microarrays, labeling, hybridization, ligation, and transformation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Kit uses glass fiber matrix technology in a 96-well format for highly efficient purification of PCR products. DNA fragments from PCR are captured by the matrix in presence of a chaotropic salt and contaminants are removed by washing matrix.
Format Kit
Isolation Technology Enzymatic PCR Clean-up Technology
Sample Type DNA Solutions, Enzyme Reactions, PCR Mixtures
For Use With (Application) Fluorescent sequencing, microarrays, labeling, hybridization, ligation and transformation
Includes Glass-fiber matrix 96-well plates binding plates, wash plates, collection plates and color-coded bottles of capture Buffer, Wash Buffer choice of two Elution Buffers (Tris HCl and sterile Water), instruction booklet
Product Type illustra GFX 96 PCR Purification Kit
Quantity 96-Well Plates
Sample Volume 100 bp-10 kbp
Yield Up to 95% when purifying a 910 bp fragment at a starting concentration of 8.4 ng/μl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.