Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GFX™ PCR DNA and Gel Band Purification Kit

Catalog No. 45001490 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
100 Preps
250 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-490 250 Preps
45-001-489 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-490 Supplier Cytiva Supplier No. 28903471
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

GFX columns; Collection tubes; Color-coded bottles of capture buffer; Wash buffer; two elution buffers: Tris-HCl and sterile water

Purify and concentrate PCR products or DNA fragments ranging in size from 50 bp to 10 kb. Cytiva Life Sciences™ illustra™ GFX™ PCR DNA and Gel Band Purification Kit can be used to purify DNA from reaction volumes up to 100μL or agarose gel slices of up to 900mg.

Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution.

  • Rapid and easy-to-use purification method with less than 10 minutes hands-on time
  • Flexible 10 to 50μL elution volume for different DNA concentration needs
  • Improved column design with increased maximum volume holding of up to 800μL liquid volume (950μL without a cap)
  • DNA of exceptional purity (to 99.5%) is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning
  • Visual color indicator added to the capture buffer to ensure optimal pH for maximum DNA binding and recovery
  • Kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin-column for the purification of DNA from both solution and agarose gel

TRUSTED_SUSTAINABILITY

Specifications

Description Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg.
Format Kit
Isolation Technology Silica Spin Column
Sample Type Agarose Gel Slices, DNA Solutions, Enzyme Reactions, PCR Mixtures
Formulation Tris HCl and sterile Water
For Use With (Application) Isolation and concentration of DNA fragments (50bp to 10kb) from PCR mixtures, DNA-containing Agarose Gel bands, Enzyme-based DNA modifications and restriction digestions
Includes GFX™ columns, collection tubes, color-coded bottles of capture Buffer, Wash Buffer, two Elution Buffers
No. of Reactions 250 Preps
Product Type illustra GFX PCR DNA and Gel Band Purification Kit
Sample Volume 50 bp-10 kb
Yield PCR 10 μl elution volume-65%, PCR 50 μl elution volume-82%, 300 mg agarose 10 μl elution volume-57%, 300 mg agarose 50 μl elution volume-91%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.