Learn More
Cytiva MicroSpin™ G-25 Columns
GSA_VA
Catalog No. 45001397 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Pack
Catalog No. Quantity
45-001-397 50 Pack
1 options
Catalog No. 45-001-397 Supplier Cytiva Supplier No. 27532501
Get an Extra Life Sciences Product with Purchase
View all promotions
Add to Cart
Add to Cart
Use for the rapid purification of DNA >10 base pairs in length in a wide range of applications. Cytiva Life Sciences™ llustra™ MicroSpin™ G-25 Columns are ideal for desalting, buffer exchange, and removal of unincorporated nucleotides from end-labeled oligonucleotides.
  • In spin-column chromatography, permit rapid buffer exchange/desalting of PCR products and other DNAs in volumes of 10 to 100μL
  • Provide rapid purification of newly synthesized oligonucleotides in 100 to 150μL of deprotection solution
  • Other applications incude desalting/buffer exchange of DNA and removal of unincorporated radionucleotides from end-labeled oligonucleotides in volumes of 10 to 100μL
  • Require less than four minutes from sample application to collection of purified product
  • Tested in nickase, single- and double-stranded exonuclease, and RNase assays
Desalting/buffer exchange of DNA, Removal of unincorporated radionucleotides from end-labeled oligonucleotides in volumes of 10 to 100μL
Order Info
Conveniently prepacked with Sephadex™ G-25 DNA Grade pre-equilibrated in distilled water containing 0.05% Kathon™ CG/ICP Biocide
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
Isolation Technology Filtration
For Use With (Application) Desalting/buffer exchange of DNA, Removal of unincorporated radionucleotides from end-labeled Oligonucleotides in volumes of 10 to 100μL
Product Type Column
Quantity 50 Pack