Learn More
Cytiva MicroSpin™ S-300 HR Columns
GSA_VA
Catalog No. 45001395 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
45-001-395 Each
1 options
This item is not returnable. View return policy
Catalog No. 45-001-395 Supplier Cytiva Supplier No. 27513001
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
For rapid purification of PCR products (>100 bp) from unincorporated primers (<20-mers) and nucleotides in a volume of 25 to 50μL using spin-column chromatography
  • Useful for purification of alkaline-denatured plasmid DNA prior to sequencing
  • Requires <4 min. from sample application to collection of purified product
  • Prepacked with Sephacryl™ S-300 HR media pre-equilibrated in TE buffer
  • Percent sample recovery: >80%
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
Isolation Technology Spin Column
For Use With (Application) Rapid purification of PCR products (>100bp) from unincorporated primers (<20-mers) and Nucleotides in a volume of 25 to 50μL using spin-column chromatography
Product Type Column
Quantity Each