Learn More
Cytiva MicroSpin™ S-400 HR Columns
Catalog No. 45001396 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
45-001-396 Each
1 options
This item is not returnable. View return policy
Catalog No. 45-001-396 Supplier Cytiva Supplier No. 27514001
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use for rapid purification of PCR products (>200 bp) from unincorporated primers (<32-mers) and nucleotides. Cytiva Life Sciences™ illustra™ MicroSpin™ S-400 HR Columns use spin-column chromatography for quick results.
  • Accommodates 25 to 50μL for post-PCR cleanup prior to cloning or a second amplification reaction; 25 to 100μL for all other applications
  • Ready to use—requires <4 min. from sample application to collection of purified product
  • Prepacked with Sephacryl™ S-400 HR media pre-equilibrated in TE buffer
  • Percent sample recovery: >80%
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays
TRUSTED_SUSTAINABILITY
Specifications
Format Spin Column
Isolation Technology Spin Column
For Use With (Application) Rapid purification of PCR products (>200bp) from unincorporated primers (<32-mers) and Nucleotides using spin-column chromatography
Product Type Column
Quantity Each