Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva NAP™ Columns, NAP-25

Catalog No. 45000150 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-150 50 Pack
45-000-149 20 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-150 Supplier Cytiva Supplier No. 17085202
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing, and sample elution

Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade

  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange

Order Info

Sephadex G-25 DNA grade gel comes in a column buffer consisting of distilled water with 0.15% Kathon™ CG/ICP Biocide.

TRUSTED_SUSTAINABILITY

Specifications

Format Column
For Use With (Application) Rapid and efficient purification of DNA and Oligonucleotides
Includes Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product Type NAP-25
Quantity 50 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.