Learn More
Cytiva NICK™ Columns
Catalog No. 45000155 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 Columns
50 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-155 20 Columns
45-000-156 50 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-000-155 Supplier Cytiva Supplier No. 17085501
Add to Cart
Add to Cart
This item is not returnable. View return policy
For rapid purification of labeled DNA (>20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography
  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0.15% Kathon™ CG/ICP Biocide
  • Typical recovery: ≥90% of applied DNA
  • Can accommodate sample volumes up to 100μL
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography
Includes Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide
Product Type Column
Quantity 20 Columns