Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva NICK™ Columns

Catalog No. 45000156 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
20 Columns
50 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-156 50 Columns
45-000-155 20 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-156 Supplier Cytiva Supplier No. 17085502
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For rapid purification of labeled DNA (>20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography

  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0.15% Kathon™ CG/ICP Biocide
  • Typical recovery: ≥90% of applied DNA
  • Can accommodate sample volumes up to 100μL

TRUSTED_SUSTAINABILITY

Specifications

Format Column
Isolation Technology Filtration
For Use With (Application) Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography
Includes Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide
Product Type Column
Quantity 50 Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.