Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva plasmidPrep Midi Flow Kit

Catalog No. 45001498 Shop All Cytiva Products
Change view
Click to view available options
PlasmidPrep Midi Flow Kit; 25 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
45-001-498 PlasmidPrep Midi Flow Kit; 25 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-498 Supplier Cytiva Supplier No. 28904267

missing translation for 'validMainframeMsg'

Includes

Fast-flow plasmid 250 columns; Resuspension, lysis, neutralization, wash, elution, and TE buffers; Full protocol booklet with a detachable, quick reference protocol card

For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification

The kit consists of a high-performance, anion-exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA.

  • High column capacity allows for a proportional yield for input sample volume of 25 to 50mL (for high copy number plasmid) and up to 500mL for low copy number plasmid
  • Provides plasmid DNA with excellent purity and low endotoxin levels
  • Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns

TRUSTED_SUSTAINABILITY

Specifications

Description illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume.
For Use With (Application) Used For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification
Includes Fast-flow genomic 250 columns, lyophilized Proteinase K powder, Lysis solutions, load and Elution Buffers, NAP-25 desalting column, spin column adapters, full protocol booklet, detachable, quick reference protocol card
No. of Reactions 25 Preps
Product Type illustra plasmidPrep Midi Flow Kit
Purity 1.8
Sample Volume 150 mL Culture
Test Time Approx.215 min
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.