Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit

Catalog No. 25660124 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
24
480
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
25-660-124 24
01-183-294 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-660-124 Supplier Cytiva Supplier No. 25660124
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Highly efficient and representative whole-genome amplification with 40 to 60μg yield from nanogram amounts of DNA sample. Cytiva Lifescience™ illustra™ Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit is part of the Phi29 DNA polymerase family of products.

p> 

  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
  • The reaction mixture is provided as predispensed, single-dose, lyophilized cakes in either strips of 8 tubes, or 96-well plates.
  • Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample.
  • Amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.
  • The average product length is over 10kb.
  • Procedure is quick, simple and does not require a thermocycler.
  • A typical DNA yield of 12 to 20μg DNA can be achieved in less than two hours from only 10ng of genomic DNA input.
  • The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or a nonpurified cell lysate may be used.
  • Protocols for the amplification of DNA from blood lysates and dried blood spots are included in the manual.
  • GenomiPhi amplified DNA is suitable for various applications such as genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival.

 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Description illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 12 to 20 μg yield from nanogram amounts of DNA sample.
Format Kit
For Use With (Application) Genomic DNA Amplification
No. of Reactions 24
Product Type illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit 24 Reactions
Size > 10 kb
Test Time 1.5-2 h
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.