Learn More
Cytiva RNAspin Mini Isolation Kit
Catalog No. 45001161 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
20 Preps
250 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-161 20 Preps
45-001-163 250 Preps
45-001-162 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-161 Supplier Cytiva Supplier No. 25050070
Add to Cart
Add to Cart
Includes
Protocol booklet with detachable, quick reference card, necessary components for specified number of preps, including prefilters and DNase I
Extract high-quality RNA from a wide range of samples rapidly. Cytiva Life Sciences™illustra™ RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast.
  • Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
  • Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
  • Prefilters reduce lysate viscosity for maximum yield and purity
  • Results can be obtained with small amounts of sample
  • Binding capacity: 100μg
  • Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
  • Elution volume: 40 to 120μL
  • RNA Purity (A260/A280): 1.8 to 2.2
  • Total time of preparation: <30 min/6 preps
Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments
TRUSTED_SUSTAINABILITY
Specifications
Description illustra RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.
For Use With (Application) Used for the isolation of high-quality total RNA from a wide range of source material including hard-to-lyse bacterial strains. Purified RNA is suitable for use in sensitive downstream applications such as quantitative reverse transcriptase PCR (qRT-PCR),
Includes Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
No. of Reactions 20 Preps
Product Type illustra RNAspin Mini Kit
RNase 1.8-2.2
Sample Volume <30 mg
Test Time < 30 min/6 preps
Yield Up to 100μg