Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva RNAspin Mini Isolation Kit
GSA_VA

Catalog No. 45001163 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
20 Preps
250 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-001-163 250 Preps
45-001-162 50 Preps
45-001-161 20 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-163 Supplier Cytiva Supplier No. 25050072
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Protocol booklet with detachable, quick reference card, necessary components for specified number of preps, including prefilters and DNase I

Extract high-quality RNA from a wide range of samples rapidly. Cytiva Life Sciences™illustra™ RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast.

  • Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
  • Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
  • Prefilters reduce lysate viscosity for maximum yield and purity
  • Results can be obtained with small amounts of sample
  • Binding capacity: 100μg
  • Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
  • Elution volume: 40 to 120μL
  • RNA Purity (A260/A280): 1.8 to 2.2
  • Total time of preparation: <30 min/6 preps

Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description illustra RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.
Format Kit
Isolation Technology Silica Membrane Technology
For Use With (Application) Used for the isolation of high-quality total RNA from a wide range of source material including hard-to-lyse bacterial strains. Purified RNA is suitable for use in sensitive downstream applications such as quantitative reverse transcriptase PCR (qRT-PCR),
Includes Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
No. of Reactions 250 Preps
Product Type illustra RNAspin Mini Kit
RNase 1.8-2.2
Sample Volume <30 mg
Test Time < 30 min/6 preps
Yield Up to 100μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.