Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Solution dNTP Sets
GSA_VA

Catalog No. 45001454 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 25 μm
4 x 100 μm
4 x 500 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-454 4 x 25 μm
45-001-455 4 x 100 μm
45-001-456 4 x 500 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-454 Supplier Cytiva Supplier No. 28406551
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Use dNTP sets for optimum performance and consistency. Cytiva Lifescience™ illustra™ Solution dNTP Sets are free from DNase, RNase, and nicking enzyme activity.

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7
Product Type dNTP set
Concentration 100 mM
Purity >99%
Quantity 4 x 25 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.