Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Solution dNTP Sets

Catalog No. 45001456 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 25 μm
4 x 100 μm
4 x 500 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-456 4 x 500 μm
45-001-454 4 x 25 μm
45-001-455 4 x 100 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-456 Supplier Cytiva Supplier No. 28406553
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use dNTP sets for optimum performance and consistency. Cytiva Lifescience™ illustra™ Solution dNTP Sets are free from DNase, RNase, and nicking enzyme activity.

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7
Product Type dNTP set
Concentration 100 mM
Purity >99%
Quantity 4 x 500 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.