Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Solution dNTPs

Use dNTPs incorporating the most recent advances in manufacturing technology. Cytiva illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

$101.20 - $321.20

Specifications

Format Liquid
pH 7
Concentration 100 mM
Purity >99%
Color Colorless
View More Specs

Products 6
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
45-001-440
View Documents
Cytiva
28406501
dATP 25 μm
Each for $101.20
Only null left
Add to Cart
 
45-001-447
View Documents
Cytiva
28406522
dGTP 100 μm
Each for $321.20
Only null left
Add to Cart
 
45-001-444
View Documents
Cytiva
28406512
dCTP 100 μm
Each for $321.20
Only null left
Add to Cart
 
45-001-450
View Documents
Cytiva
28406532
dTTP 100 μm
Each for $321.20
Only null left
Add to Cart
 
45-001-452
View Documents
Cytiva
28406541
dUTP 25 μm
Each for $101.64
Only null left
Add to Cart
 
45-001-441
View Documents
Cytiva
28406502
dATP 100 μm
Each for $321.20
Only null left
Add to Cart
 
Description

Description

 

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

 

Order Info

Custom formulation, testing, packaging, and labeling are available

Specifications

Specifications

Liquid
100 mM
Colorless
7
>99%
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.