Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva TempliPhi™ 100/500 Amplification Kit
GSA_VA

Catalog No. 45002265 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-002-265 500
45-002-262 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-265 Supplier Cytiva Supplier No. 25640050
Get an Extra Life Sciences Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Sample buffer, Reaction buffer, Enzyme mix, and Positive control pUC19 DNA (2ng/μL)

Use amplified DNA directly for cycle sequencing without purification. Cytiva illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.

  • Generate microgram quantities of template DNA from picogram amounts of starting material
  • Completed reactions contain uniform quantities of DNA irrespective of input amount, allowing for use directly in sequencing reactions without quantification
  • Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA without the need for purification
  • Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
  • Contain separate, unmixed components for enhanced stability

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The product can be stored for 2 months or less at -20°C. For storage longer than 2 months, store at -70°CThaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification
Description illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.
Format Kit
For Use With (Application) Sequencing Template Amplification Kits and Reagents
Includes 0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20μL Enzyme mix and 50μL positive control pUC19 DNA
No. of Reactions 500
Product Type illustra TempliPhi 500 Amplification Kit
Test Time 4-18 h depending on starting material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.