Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva TempliPhi™ 2000 Reaction Kit
GSA_VA

Catalog No. 45003001 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
45-003-001 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-003-001 Supplier Cytiva Supplier No. 28964286
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

TempliPhi Premix, Denature buffer, Enzyme mix, and Positive control pUC19 DNA (2ng/μL)

Uses isothermal rolling circle amplification (RCA) to efficiently prepare DNA sequencing templates with a formulation designed for higher throughput applications

  • Simple protocol reduces time, labor and consumables needed for template preparation, and workflow enables easy automation
  • Generate microgram quantities of template DNA from picogram amounts of starting material
  • Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks or purified circular (plasmid or M13) DNA
  • Use amplified DNA directly for cycle sequencing without purification
  • Prepare circular DNA templates for cycle sequencing, cloning, and transformation in 4 to 6 h

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Denature buffer and control DNA at -70°C or -15°C to -30°C. Store TempliPhi premix at -70°C.Do not store TempliPhi premix at -15°C to -30°C. Thaw the premix on ice and maintain at 0–4°C during handling. Do not allow premix to warm above 4°C prior
Description illustra TempliPhi 2000 Reaction Kit
Format Kit
For Use With (Application) High-throughput preparation of circular DNA templates for cycle sequencing, cloning, and transformation.
Includes TempliPhi™ premix, denature Buffer, Enzyme mix and positive control pUC19 DNA
No. of Reactions 2000
Product Type illustra TempliPhi 2000 Reaction Kit
Test Time 16-18 h
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.