Learn More
Sigma Aldrich Fine Chemicals Biosciences illustraTM PuReTaq RTG PCR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE27955702
Encompass
PuReTaq Ready-To-Go PCR Beads are pre-mixed predispensed single-dose reactions optimized for performing standard PCR amplifications. The use of recombinant PuReTaq DNA polymerase and other high-purity reagents ensures reliable and robust performance in both end point and real-time fluorescence-based PCR amplifications and ensures the lowest possible levels of contaminating prokaryotic and eukaryotic nucleic acids.The puReTaq Ready-To-Go PCR Beads are pre-formulated to ensure greater reproducibility between reactions minimize pipetting steps and reduce the potential for pipetting errors and contamination. The only additional reagents required are water primers and template DNA. The beads are provided pre-dispensed into either 0.2 ml or 0.5 ml PCR tubes. The 0.2 ml tubes are also supplied in a 96-well (8 x 12) plate format that allows individual strips of eight tubes to be easily removed. This flexibility allows use of either the entire 96-well plate strips of eight or individua
Catalog No. 50-290-8310
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.