Learn More
ILVBL Mouse anti-Human, Clone: OTI3E9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503141

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.Specifications
ILVBL | |
Monoclonal | |
Unconjugated | |
ILVBL | |
209L8, AHAS, HACL1L, ILV2H | |
Mouse | |
Affinity Chromatography | |
RUO | |
10994 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
A1L0T0 | |
ILVBL | |
Full length human recombit protein of human ILVBL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.