Learn More
Enzo Life Sciences IM-54 (1mg). CAS: 861891-50-1
SDP
Supplier:  Enzo Life Sciences ALX430137M001
Encompass_Preferred
Cell permeable inhibitor of necrosis. Selectively blocks oxidative stress-induced necrotic cell death (~3µM IM-54 prevented ~50% cell death in HL60 cells exposed to 100µM H2O2). Does not protect against etoposide-induced apoptosis or display antioxidant properties. Does not affect S6K1 and PKC kinase activities at concentrations of 50µM. Alternative name: 2-(1H-indol-3-yl)-3-pentylamino-maleimide. Purity: ≥98% (HPLC). Appearance: Yellow powder. Solubility: Soluble in DMSO (100mg/ml), dichloromethane, 100% ethanol or methanol (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9227
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.