Learn More
Medchemexpress LLC HY-136093A 10mg , IM156 CAS:1422365-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136093A10MG
Encompass_Preferred
HY-136093A 10mg IM156 CAS: 1422365-94-3 IM156 (HL156A; HL271 acetate), a chemical derivative of Metformin (HY-B0627), is a potent and orally active AMPK activator that increases AMPK phosphorylation. IM156 attenuates aging-associated cognitive impairment in animal model. IM156 is a potent oxidative phosphorylation (OXPHOS) inhibitor which can be used for the research of solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0780
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.