Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iMAC2, Tocris Bioscience™

Catalog No. 379450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-945-0 50 mg
37-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-945-0 Supplier R&D Systems Supplier No. 3794/50
This item is not returnable. View return policy

Suppressor of mitochondrial apoptosis

Inhibitor of mitochondrial apoptosis-induced channel (MAC) (IC50 = 28 nM). Reduces STS-induced apoptosis in FL5.12 cells by over 50%. Inhibits release of cytochrome c by Bid-induced Bax activation (IC50 = 0.68 μM).

Chemical Identifiers

CAS 335166-00-2
Molecular Formula C19H22Br2Cl2FN3
Molecular Weight (g/mol) 542.112
InChI Key JWCVUOLQOHIRML-UHFFFAOYSA-N
Synonym imac2, 3,6-dibromo-9-2-fluoro-3-1-piperazinyl propyl-9h-carbazole dihydrochloride, 3,6-dibromo-9-2-fluoro-3-piperazin-1-yl propyl carbazole dihydrochloride
PubChem CID 90488878
IUPAC Name 3,6-dibromo-9-(2-fluoro-3-piperazin-1-ylpropyl)carbazole;dihydrochloride
SMILES C1CN(CCN1)CC(CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)F.Cl.Cl

Specifications

Quantity 50 mg
Formula Weight 542.11
Percent Purity >98%
Chemical Name or Material iMAC2
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.