Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Image-iT™ Fixation/Permeabilization Kit

Catalog No. A5818101
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5818101 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5818101 Supplier Invitrogen™ Supplier No. A5818101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Image-iT Fixation/Permeabilization Kit is a convenient-to-use fixation/permeabilization kit that contains all the necessary reagents to prepare cells for antibody staining and imaging.

The Image-iT Fixation/Permeabilization Kit is a convenient-to-use fixation/permeabilization kit that contains all the necessary reagents to prepare cells for antibody staining and imaging. The high-quality components help preserve cell morphology and reduce background staining and are ideal for use with antibody-based detection methods, including immunocytochemistry (ICC), immunofluorescence (IF), and immunohistochemistry (IHC). An easy-to-follow protocol is also provided. Note: the 4% formaldehyde, methanol-free fixative solution included in this kit is also available for separate purchase (see Cat. No. R37814).

Features of the Image-iT Fixation/Permeabilization Kit include:
• A complete fixation/permeabilization kit with ready-to-use components and room temperature storage
• Optimized for secondary antibody labeling, fixed-cell dye staining, and GFP applications
• Reduced background staining

The Image-iT Fixation/Permeabilization Kit contains:
Fixative: high purity 4% formaldehyde (methanol-free) in PBS, pH 7.3
Permeabilization solution: 0.5% Triton X-100
Blocking buffer: 3% BSA, fraction V, de-lipidated, New Zealand source, in DPBS
Wash solution: DPBS, no calcium, no magnesium

TRUSTED_SUSTAINABILITY

Specifications

Product Line Image-iT
Content And Storage • Image-iT Fixative solution
• Permeabilization solution
• Blocking solution
• Wash solution DPBS

Store at room temperature.
Quantity 1 Kit
Shipping Condition Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.