Learn More
Agilent BioTek Imaging Objectives for Lionheart FX Automated Live Cell Imager
Shop All Agilent Technologies Products
Description
- Includes oil and absorbing ring
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | 100X Oil Objective, 1.40 NA |
| For Use With (Equipment) | Lionheart FX Automated Live Cell Imager |