Learn More
STEMCELL Technologies Imatinib (Mesylate), Size: 25 mg
SDP
Supplier:  STEMCELL Technologies 72532
Encompass_Preferred
Imatinib mesylate is a first generation tyrosine kinase inhibitor that selectively targets certain tyrosine kinases, including ABL, platelet-derived growth factor receptor (PDGFR), and KIT (Druker, 2008; Müller).DIFFERENTIATION· Inhibits proliferation of primary cultured human mesenchymal stem cells, and promotes adipogenic over osteogenic differentiation (Fierro et al.).· Induces osteoblast differentiation in cultured osteoblastic cells, and reduces osteoclastogenesis in mouse bone marrow cultures (O’Sullivan et al.). CANCER RESEARCH· In CML, Imatinib inhibits the oncoprotein BCR-ABL, the product of the Philadelphia chromosome gene fusion (Carroll et al.; Druker et al., 1996).· Inhibits autonomous erythropoiesis in peripheral blood mononuclear cells isolated from patients with polycythemia vera (Oehler et al.).
Catalog No. 50-197-5937
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.