Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imatinib Mesylate 98.0+%, TCI America™
SDP

Catalog No. I0936100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0936100MG 100 mg
I09361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0936100MG Supplier TCI America Supplier No. I0936100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 220127-57-1
Molecular Formula C30H35N7O4S
Molecular Weight (g/mol) 589.72
MDL Number MFCD04307699
InChI Key YLMAHDNUQAMNNX-UHFFFAOYSA-N
Synonym imatinib mesylate, gleevec, glivec, imatinib mesilate, imatinib methanesulfonate, imatinib mesylate sti571, imatinib monomesylate, unii-8a1o1m485b, imatinib, methanesulfonate salt
PubChem CID 123596
ChEBI CHEBI:31690
IUPAC Name N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide; methanesulfonic acid
SMILES CS(O)(=O)=O.CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 589.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imatinib Mesylate
RTECSNumber CV5520550
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.