Learn More
Selleck Chemical LLC Imatinib (STI571) 152459-95-5 Gleevec, Glivec, CGP057148B
SDP
Supplier:  Selleck Chemical LLC S247510MM/1ML
Encompass_Preferred
Imatinib (STI571, CGP057148B, Gleevec) is a multi-target inhibitor of tyrosine kinase with inhibition for v-Abl, c-Kit and PDGFR, IC50 values are 0.6 ?M, 0.1 ?M and 0.1 ?M in cell-free or cell-based assays, respectively. Imatinib (STI571) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1893466
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.