Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Imb-301 | 64009-84-3 | 99.1% | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1221565MG

Encompass_Preferred

Imb-301 is a small-molecule inhibitor of HIV-1 replication that binds human APOBEC3G (hA3G), disrupts the hA3G-Vif interaction, and inhibits Vif-mediated degradation of hA3G. It is supplied as a research reagent and intended for laboratory use only.

  • Inhibits Vif-mediated degradation of hA3G.
  • High purity (99.1%).
  • Molecular weight 379.26 g/mol.
  • CAS number 64009-84-3 for unambiguous identification.
  • Supplied as an oil in a 5 MG quantity for laboratory use only.

Catalog No. 50-002-30620


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.