Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iMDK, Tocris Bioscience™
GREENER_CHOICESDP

Catalog No. 512650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-265-0 50 mg
51-261-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-265-0 Supplier Novus Biologicals Supplier No. 5126/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Suppressor of MDK expression; inhibits PI 3-K signaling

Suppressor of endogenous midkine (MDK) expression. Inhibits PI 3-K signaling in H441 lung adenocarcinoma cells. Selectively attenuates growth of MDK-expressing cancer cells. Induces apoptosis in H441 cells and suppresses growth of H441 cell xenografts in mice.

Chemical Identifiers

CAS 881970-80-5
Molecular Formula C21H13FN2O2S
Molecular Weight (g/mol) 376.405
InChI Key IWFKQTWYILKFGE-UHFFFAOYSA-N
Synonym imdk, 3-2-4-fluorobenzyl imidazo 2,1-b 1,3 thiazol-6-yl-2h-chromen-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b thiazol-6-yl-2h-1-benzopyran-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b 1,3 thiazol-6-yl chromen-2-one, 3-2-4-fluorophenyl methyl imidazo 2,1-b 1,3-thiazolin-6-yl chromen-2-one
PubChem CID 15991416
IUPAC Name 3-[2-[(4-fluorophenyl)methyl]imidazo[2,1-b][1,3]thiazol-6-yl]chromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C3=CN4C=C(SC4=N3)CC5=CC=C(C=C5)F

Specifications

Quantity 50 mg
Formula Weight 376.4
Chemical Name or Material iMDK
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.